1-Propanol 99% Spectrophotometry

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane

22933.K2
22933.K2EA 148 CHF
22933.K2
1-Propanol 99% Spectrophotometry
1-Propanol

Purity: 99% CAS No.: 71-23-8 Molecular Formula: CH3CH2CH2OH Formula Weight: 60.10 Flash Point: 15°(59°F) Physical Form: Liquid Density: 0.804 Refractive Index: 1.3850 Odour: Alcohol-like Melting Point: -127° Boiling Point: 97° Beilstein Reference: 1098242 Merck Reference: 14,7842 UN No.: 1274 MDL No.: MFCD00002941

Formel: H₃CCH₂CH₂OH
MW: 60.1 g/mol
Siedepunkt: 97 °C
Melting Pt: –127 °C
Dichte: 0.804
Flash-Pt: 15 °C (59 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00002941
CAS-Nummer: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3 II
Merck Index: 14,7842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR