Tubing holder for beakers, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F182350000
229-0001EA 75.2 CHF
229-0001
Tubing holder for beakers, SP Bel-Art
Klemmen Schlauchklemmen und Sanitärklemmen
This PP clip holds flexible tubing steady and fits securely onto the edge of beakers or pitchers up to ¼" thick.
  • 5,1 cm (2") long arm has a hole to accept tubing up to 9,5 mm (⅜") Ext. Ø
  • Autoclavable