Chloroform 99,0-99,6 Stabilised, GPR RECTAPUR®

Lieferant: VWR Chemicals
Gefahr

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilization: 0.6 % EtOH

22706.292EA 60.1 CHF
22706.292 22706.326 22706.361 22706.463 22706.554
Chloroform 99,0-99,6 Stabilised, GPR RECTAPUR®
Chloroform
Formel: CHCl₃
MW: 119.38 g/mol
Siedepunkt: 61.7 °C (1013 hPa)
Melting Pt: –63 °C
Dichte: 1.4832 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00000826
CAS-Nummer: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1 III
REACH: 01-2119486657-20
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Assay 99.0 - 99.6 %
Boiling point 59.5 - 60.5 °C
Density (20/4) 1.473 - 1.489
E/E 0.1 - 1.0 %
Evaporation residue Max. 50 ppm
Free chlorine Max. 10 ppm
Heavy metals (as Pb) Max. 10 ppm
Water Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR