Bariumaluminat, tech.

Lieferant: Thermo Fisher Scientific
Achtung

22642.22 22642.36
22642.22EA 120 CHF
22642.22 22642.36
Bariumaluminat, tech.
Barium aluminate

CAS No.: 12004-04-5 Molecular Formula: BaAl2O4 (approximate formula) Formula Weight: 561.98 Physical Form: Powder Odour: Odourless UN No.: 1564 MDL No.: MFCD00049716

Formel: Al₂BaO₄
Melting Pt: 169.5…170 °C
Dichte: 4.04 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00049716
CAS-Nummer: 12004-04-5
EINECS: 234-445-9
UN: 1564
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR