Kaliumiodid ≥99,0% ACS, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

Synonyms: Dipotassium diiodide, Kali iodide

221945-100G 221945-2.5KG 221945-500G 221945-12KG 221945-25KG
221945-100GEA 88.84 CHF
221945-100G 221945-2.5KG 221945-500G 221945-12KG HONC221945-25KG
Kaliumiodid ≥99,0% ACS, Fluka™
Kaliumiodid
Formel: Kl
Siedepunkt: 1.323 °C (1.013 hPa)
Melting Pt: 681 °C
Dichte: 3.12 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011405
CAS-Nummer: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now

Spezifikation Testergebnisse

Assay Min. 99.0 %
pH (5 %, 25 °C) 6.0 - 9.2
Loss on drying Max. 0.2 %
Water insoluble matter Max. 0.005 %
Barium (Ba) Max. 0.002 %
Calcium (Ca) Max. 0.002 %
Iron (Fe) Max. 3 ppm
Magnesium (Mg) Max. 0.001 %
Sodium (Na) Max. 0.005 %
Heavy metals (as Pb) Max. 5 ppm
Bromide, chloride (as Cl) Max. 0.01 %
Iodate (IO₃) Max. 3 ppm
Phosphate (PO₄) Max. 0.001 %
Sulfate (SO₄) Max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR