Lithiumchlorid Monohydrat ≥99,95% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Lithium chloride hydrate, Hydrochloric acid lithium salt monohydrate

22151.14 22151.22
22151.14EA 294 CHF
22151.14 22151.22
Lithiumchlorid Monohydrat ≥99,95% (Metall-Basis)
Lithiumchlorid Monohydrat

Purity: 99.95% CAS No.: 16712-20-2 Molecular Formula: LiCl.H2O Formula Weight: 60.41 (42.39anhy) Physical Form: Crystalline Density: 1.78 Melting Point: 98° -H2O Merck Reference: 14,5528 MDL No.: MFCD00011078

Formel: ClLi·1H₂O
MW: 60.41 g/mol
Melting Pt: >98 °C (-H₂O)
Dichte: 1.78
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149764
CAS-Nummer: 16712-20-2
EINECS: 231-212-3
Merck Index: 14,5528

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR