Paraformaldehyd 4% in PBS anwendungsbereit, fixatives Reagens

Lieferant: Biotium
Gefahr

Synonyms: Polyoxymethylene

Packaged under argon.

22023
22023.EA 278 CHF
22023.
Paraformaldehyd 4% in PBS anwendungsbereit, fixatives Reagens
Paraformaldehyd

Paraformaldehyde, 4% in PBS is a ready-to-use fixation solution for cells or tissues.


  • Prepared from EM grade paraformaldehyde
  • No glass ampoules to break, store in the original bottle
  • Safer and more convenient than handling paraformaldehyde solid


It is electron microscopy-grade paraformaldehyde dissolved in pH 7,4 PBS, and is stabilised by packaging under argon with no methanol added.

Formel: (CH₂O)n
MDL Number: MFCD00133991
CAS-Nummer: 30525-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR