Wasserstoffperoxid 30%, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Gefahr

2192.0250EA 41.6 CHF
2192.0250 2192.1000 2192.2500PE 2192-39
Wasserstoffperoxid 30%, BAKER ANALYZED® ACS, J.T.Baker®
Wasserstoffperoxid
Formel: H₂O₂
MW: 34.01 g/mol
Siedepunkt: ∼107 °C (1013 hPa)
Melting Pt: ≤ 0 °C
Dichte: 1.11 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011333
CAS-Nummer: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1 II
Merck Index: 14,04798

Order Now

Spezifikation Testergebnisse

Exceeds ACS Specifications
Assay 30.0 - 32.0 %
Ammonium (NH4) max. 5 ppm
Color (APHA) max. 10
Chloride (Cl) max. 1 ppm
Heavy Metals (as Pb) max. 1 ppm
Nitrate (NO3) max. 2 ppm
Phosphate (PO4) max. 2 ppm
Residue after Evaporation max. 0.002 %
Sulfate (SO4) max. 2 ppm
Titrable Acid (meq/g) max. 0.0006
Trace Impurities (in ppm)
Silver (Ag) max. 0.01
Aluminium (Al) max. 0.05
Arsenic (As) max. 0.05
Barium (Ba) max. 0.02
Beryllium (Be) max. 0.01
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.02
Cobalt (Co) max. 0.01
Chromium (Cr) max. 0.1
Copper (Cu) max. 0.01
Iron (Fe) max. 0.1
Germanium (Ge) max. 0.05
Potassium (K) max. 0.1
Lithium (Li) max. 0.01
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.01
Molybdenum (Mo) max. 0.02
Sodium (Na) max. 0.5
Nickel (Ni) max. 0.05
Lead (Pb) max. 0.05
Strontium (Sr) max. 0.01
Titanium (Ti) max. 0.1
Thallium (Tl) max. 0.05
Vanadium (V) max. 0.01
Zinc (Zn) max. 0.05
Zirconium (Zr) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR