Karbolfuchsin (Ziehl-Neelsen) zur Mikroskopie und Bakteriologie phenolfrei, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyms: 4-[(4-Amino-m-tolyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]-o-toluidine hydrochloride

M-Clarity™ Qualitätsstufe = MQ100

21820-100ML 21820-1L
21820-100MLEA 36.7 CHF
21820-100ML 21820-1L
Karbolfuchsin (Ziehl-Neelsen) zur Mikroskopie und Bakteriologie phenolfrei, Sigma-Aldrich®
Karbolfuchsin

It is used as diagnostic assay manufacturing, hematology, histology.

Formel: C₂₀H₂₀ClN₃
MW: 351.88 g/mol
Dichte: 0.99 g/cm³ (20 °C)
Lager Temperatur: Room Temperature
MDL Number: MFCD18071399
CAS-Nummer: 632-99-5
EINECS: 211-189-6
UN: 1992
ADR: 3 III

Order Now

Spezifikation Testergebnisse

Appearance (colour) Red to very dark red
Appearance (Form) Liquid
Density 0.987 - 0.993
Wavelength (1)(UV) 542 - 552 nm
Extinction (1)(UV) 1%/1 cm
Solvent Ethanol/H₂0 1/1

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR