Vierkant-Weithalsflaschen mit Schraubverschluss, Nalgene®
Lieferant: Thermo Fisher Scientific
2110-0002 2110-0006 2110-0008 2110-0016 2110-0032 2114-0002 2114-0006 2114-0008 2114-0016 2114-0032
215-7272EA 48.5 CHF
215-7272 215-7273 215-7274 215-7275 215-7276 215-7282 215-7283 215-7284 215-7285 215-7286
Vierkant-Weithalsflaschen mit Schraubverschluss, Nalgene®
Flaschen Kunststoffflaschen
HDPE, translucent or PPCO, translucent, with PP screw cap.
  • HDPE: Compatibility with most alcohols, bases and acids. PPCO: Very good chemical resistance
  • HDPE: Heat resistant up to +120 °C. PPCO: Heat resistant up to +121 °C, autoclavable
  • Leakproof
Ideal for storing and shipping samples. Wide mouth permits easy filling and cleaning. Square shape saves space.