Carboys, with handles and stopcock, HDPE
Lieferant: SEMADENI
1014
215-0290EA 180 CHF
215-0290
Carboys, with handles and stopcock, HDPE
Flaschen Ballons
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
With stopcock, black screw cap and black handle.