Erlenmeyer flasks, narrow neck, with baffles
Lieferant: NEUBERT VOLUME GLASSWAERE
0749-10-0250 0749-10-0500 0749-10-1000 0749-10-2000 0749-30-0250 0749-30-0500 0749-30-1000 0749-30-2000 0749-10-0100 0749-10-5000 0749-30-0100
214-0182EA 126 CHF
214-0182 NEUB0749-10-0500 214-0183 214-0184 214-0185 214-0186 214-0187 NEUB0749-30-2000 NEUB0749-10-0100 NEUB0749-10-5000 NEUB0749-30-0100
Erlenmeyer flasks, narrow neck, with baffles
Kolben Erlenmeyerkolben
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Ideal for use in the preparation of cultures.