Calciumhydroxid ≥96,0% (durch titrimetrische Analyse), Purum p.a.

Lieferant: Honeywell Chemicals
Gefahr

Synonyms: Slaked lime, Calcium dihydroxide

21181-1KG 21181-5KG 21181-25KG
21181-1KGEA 101.05 CHF
21181-1KG 21181-5KG HONC21181-25KG
Calciumhydroxid ≥96,0% (durch titrimetrische Analyse), Purum p.a.
Calciumhydroxid

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formel: Ca(OH)₂
Siedepunkt: Decomposes on heating
Melting Pt: 580 °C
Dichte: 2.24 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00010901
CAS-Nummer: 1305-62-0
EINECS: 215-137-3
ADR: 8,III
Merck Index: 13,01675

Order Now

Spezifikation Testergebnisse

Assay 96.0 - 102.0 %
Carbonate (as CaCO₃) Max. 4 %
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Copper (Cu) Max. 50 ppm
Iron (Fe) Max. 500 ppm
Potassium (K) Max. 200 ppm
Sodium (Na) Max. 100 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 50 ppm
Total S (as SO₄) Max. 500 ppm
Chloride (Cl) Max. 200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR