Racks for microcentrifuge tubes, No-Wire™, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F188380015
211-7611EA 79.7 CHF
211-7611
Racks for microcentrifuge tubes, No-Wire™, SP Bel-Art
Gestelle Reagenzglasgestelle
These PP racks are available in different grid sizes that are with open sides which make them adaptable for use in a tray of crushed ice.
  • Very good chemical resistance
  • Very good temperature resistance