Dulbecco's phosphate-buffered saline (DPBS), Corning®
Lieferant: Corning
21-031-CM 55-031-PB 55-031-PC 20-030-CV 20-031-CV 21-030-CV 21-031-LX 21-031-LB 21-031-CV 21-031-CM 21-030-CM
MDTH21-031-CMEA 142 CHF
MDTH21-031-CM 55-031-PB 55-031-PC 20-030-CV 20-031-CV 21-030-CV 21-031-LX 21-031-LB 21-031-CV 21-031-CM 21-030-CM
Dulbecco's phosphate-buffered saline (DPBS), Corning®
Puffer Gewebekultur-Puffer
Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7,0 to 7,6
  • Low endotoxin - < 0,25 EU/ml
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.
Zertifizierungen: Complete Certificate of Analysis available for each production lot, along with full formulation information.
Bitte beachten Storage temperature: Solutions 2 - 30 °C; Powder formats 2 - 8 °C
Order Now
TECHNISCHE DATEN
Environmentally Preferable
Endotoxingehalt