Isobutylmagnesium chloride 2 M diethyl ether, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

209521000
209521000.EA 70.2 CHF
209521000.
Isobutylmagnesium chloride 2 M diethyl ether, AcroSeal®
Isobutylmagnesium chloride

Appearance: Grey to yellow-brown Solution

Formel: C₄H₉ClMg
MW: 116.871 g/mol
Dichte: 0.83 g/cm³
Flash-Pt: –040 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00000471
CAS-Nummer: 5674-02-2
UN: 3399
ADR: 4.3 I

Order Now

Spezifikation Testergebnisse

Appearance Grey to yellow-brown Solution
Titration Grignard 2 M Exact strength on the certificate of analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR