Cadmium ionophore I, Selectophore™ Ionophor, Supelco®

Lieferant: Merck

Synonyms: ETH 1062, 2,2'-(Ethane-1,2-diylbis(oxy))bis(N,N-dibutylethanethioamide), N,N,N′,N′-Tetrabutyl-3,6-dioxaoctanedi(thioamide)

M-Clarity™ quality level = MQ100

20909-50MG-F
20909-50MG-FEA 583 CHF
20909-50MG-F
Cadmium ionophore I, Selectophore™ Ionophor, Supelco®
Cadmium ionophore I

Selective ionophore for cadmium used in solvent polymeric membranes.


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₂₂H₄₄N₂O₂S₂
MDL Number: MFCD00143893
CAS-Nummer: 73487-00-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d53c4eda96a619e5fb6f81096600d525dc45fe3&vid=f2afc877d6f59c43101d07c9c08eae7b6b96b692&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR