Methanol ≥99,8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., Gradienten-Qualität für die HPLC

Lieferant: VWR Chemicals
Gefahr

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

Filtered through a 0.2 µm filter, packaged under nitrogen.

20864.320EA 115 CHF
20864.320 20864.420 20864.290 20864.360 20864.400 20864.910
Methanol ≥99,8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., Gradienten-Qualität für die HPLC
Methanol
Formel: CH₃OH
Siedepunkt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Dichte: 0.79 g/cm³ (20 °C)
Flash-Pt: 11 °C (closed cup)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00004595
CAS-Nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1),II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Assay (on Anhydrous Substance) Min. 99.8%
Acidity Max. 0.0003 meq/g
Alkalinity Max. 0.0002 meq/g
Boiling point 64 - 65 °C
Density (20/20) 0.791 - 0.793
Evaporation Residue Max. 5 ppm
Water Max. 0.03%
Absorbance (210 nm) Max. 0.7
Absorbance (220 nm) Max. 0.3
Absorbance (230 nm) Max. 0.15
Absorbance (235 nm) Max. 0.1
Absorbance (250 nm) Max. 0.02
Absorbance (260 nm) Max. 0.01
Transmittance (210 nm) Min. 20%
Transmittance (220 nm) Min. 50%
Transmittance (225 nm) Min. 68%
Transmittance (230 nm) Min. 74%
Transmittance (235 nm) Min. 80%
Transmittance (250 nm) Min. 95%
Transmittance (260 nm) Min. 98%
Transmittance (260 - 420 nm) Min. 98%
Fluorescence (as Quinine; 254 nm) Max. 1 ppb
Conforms Ph. Eur. R 1053200 Passes test
Conforms Ph. Eur. R1 1053201 Passes test
Conforms Ph. Eur. R2 1053202 Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR