Wasserstoffperoxid 35%, INTEROX®

Lieferant: SOLVAY
Gefahr

206976 206978
206976.EA 270 CHF
206976. 206978.
Wasserstoffperoxid 35%, INTEROX®
Wasserstoffperoxid

INTEROX® SG-35-Plus Hydrogen peroxide 35% is a specially designed grade of hydrogen peroxide registered as biocidal product under the Reg.EU 528/2012, for professional uses in PT2 applications in the pharmaceutical, medicinal and industrilal sector for the disinfection of the surfaces of rooms, isolators and confined spaces.


Zertifizierungen: BRP authorisation n° EU-0027468-0004 1-3

Formel: H₂O₂
Siedepunkt: 107,4 °C
Dichte: 0,8835
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011333
CAS-Nummer: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1 (8),II

Order Now

Spezifikation Testergebnisse

Assay (GC) 35,0-35,7%
stability (1h,96°) ≤ 3g/kg*h
non-volatile matter (105°) <2ppm
calcium ≤0,05 ppm
iron ≤0,01 ppm
aluminium ≤0,05 ppm
sodium ≤0,5 ppm
chloride ≤0,05 ppm
phosphate (PO43-) ≤1 ppm
sulfate ≤0,5 ppm
cadmium ≤0,002 ppm
chromium ≤0,002 ppm
cobalt ≤0,002 ppm
copper ≤0,002 ppm
manganese ≤0,002 ppm
magnesium ≤0,01 ppm
nickel ≤0,002 ppm
tin ≤0,002 ppm
zinc ≤0,002 ppm
silicon ≤0,01 ppm
nitrate ≤3 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR