Anti-IgG Donkey Polyclonal Antibody (CF™ 405S)
Lieferant: Biotium
20419-1MG
20419-1MGEA 441 CHF
20419-1MG
Anti-IgG Donkey Polyclonal Antibody (CF™ 405S)
Antikörper
This product is prepared by labeling highly cross-adsorbed donkey anti-rat IgG (H+L) with CF™405S. CF™405S is a blue fluorescent dye with an absorption peak wavelength that nearly coincides with the 405 nm blue diode laser line. In addition, the emission peak wavelength of the dye well centers within the blue detection window of BD flow cytometers. As a result, flow cytometry analysis using CF™405S dye results in much brighter signal than that using Alexa Fluor® 405.   To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamster,More Product Information