EDTA Dinatriumsalz Dihydrat 99,0-101,0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens

Lieferant: VWR Chemicals

Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate

20302.236EA 110 CHF
20302.236 20302.293 20302.180 20302.464 20302.260
EDTA Dinatriumsalz Dihydrat 99,0-101,0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens
EDTA Dinatriumsalz Dihydrat
Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Melting Pt: 255
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00150037
CAS-Nummer: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Assay 99.0 - 101.0 %
pH (25°C; 5 %) 4.0 - 5.5
Identification (IR) Passes test
Identification B Passes test Ph.Eur.
Identification (Na) Passes test
Appearance of Solution S Passes test Ph.Eur.
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 50 ppm
Loss on drying (150°C) 9 - 10 %
Nitrilotriacetic acid Max. 0.05 %
Cl (Chloride) Max. 40 ppm
CN (Cyanide) Max. 10 ppm
SO4 (Sulphate) Max. 0.02 %
Cu (Copper) Max. 10 ppm
Fe (Iron) Max. 5 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR