Anti-HA tag Rabbit Polyclonal Antibody (CF®640R)

Lieferant: Biotium
20240
20240.EA 674 CHF
20240.
Anti-HA tag Rabbit Polyclonal Antibody (CF®640R)
Antikörper
Type: Tag
Antigen: HA tag (HA tag)
Clonality: polyclonal
Clone:
Conjugation: CF® 640R Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR