Biological indicators, self-contained tests, STERILtest
Lieferant: LIOFILCHEM
91100 91101 20199 91150
91100.EA 829 CHF
91100. 91101. 20199-VWR 91150.
Biological indicators, self-contained tests, STERILtest
Sterilisationsindikatoren
Biological indicators for validating the sterilisation using steam sterilisation (autoclaves).
Devices with nutrient medium, pH indicator and discs with spores of Geobacillus stearothermophilus (GST) for steam sterilisation or Bacillus atropheus (BAT) for ethylene oxide sterilisation.
Order Now
TECHNISCHE DATEN
Environmentally Preferable