Multiple adapters
Lieferant: Lenz Laborglas GmbH & CO.KG
05005111 05005444
271-2221EA 69.3 CHF
271-2221 201-2217
Multiple adapters
Destillationsbauteile Mehrhalsaufsätze
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With one standard ground NS14/23 or NS 29/32 socket and two standard ground NS 14/23 or NS 29/32 cons. Side neck descending angle of 75°.
Order Now
TECHNISCHE DATEN
Environmentally Preferable