Ethyl eosin 95 Dye, certified_Biological Stain Commission, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyms: 2',4',5',7'-Tetrabromoeosin ethyl ester potassium salt, Eosin alcohol soluble, Solvent Red 45

MQ200

199540-25G 199540-5G
199540-25GEA 401 CHF
199540-25G 199540-5G
Ethyl eosin 95 Dye, certified_Biological Stain Commission, Sigma-Aldrich®
Ethyl eosin

Certified for use in the Stoval and Black method for demonstrating Negri bodies in the central nervous system of rabid animals.

Formel: C₂₂H₁₁Br₄KO₅
MW: 714.03 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00005039
CAS-Nummer: 6359-05-3

Order Now

Spezifikation Testergebnisse

λmax 532 nm

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR