Brilliant black BN

Lieferant: Thermo Fisher Scientific

Synonyms: Food Black 1, Brilliant Black A, C.I. 28440, Black PN, Blue Black BN

199290050 199290250
199290050.EA 23.2 CHF
199290050. 199290250.
Brilliant black BN
Brilliant black BN
Formel: C₂₈H₁₇N₅O₁₄S₄
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00078980
CAS-Nummer: 2519-30-4
EINECS: 219-746-5

Order Now

Spezifikation Testergebnisse

Appearance black powder
Infrared spectrum Conforms
UV Lambda max 567 to 573 nm
Specific extinction E (1%/1cm) ≥25000 (15 mg/L in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR