Kupfer(II)acetat, wasserfrei

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Acetic acid copper (II) salt, Cupric acetate, Copper diacetate

19417.14 19417.22 19417.36
19417.14EA 201 CHF
19417.14 19417.22 19417.36
Kupfer(II)acetat, wasserfrei
Kupfer(II)acetat

CAS No.: 142-71-2 Molecular Formula: Cu(OOCCH3)2 Formula Weight: 181.63 Physical Form: Powder Sensitivity: Hygroscopic Appearance: Blue-green Odour: Odourless Merck Reference: 14,2624 UN No.: 3077 MDL No.: MFCD00008690

Formel: CuCH₃CO₂
MW: 181.64 g/mol
Siedepunkt: 240
Melting Pt: 115
Dichte: 1.9 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008690
CAS-Nummer: 142-71-2
EINECS: 205-553-3
UN: 3077
ADR: 9 III
Merck Index: 14,2624

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR