Diisobutylaluminiumhydrid 1 M hexane, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Di-i-butylaluminum hydride, DIBAL-H

183794000 183791000 183798000
183794000.EA 191 CHF
183794000. 183791000. 183798000.
Diisobutylaluminiumhydrid 1 M hexane, AcroSeal®
Diisobutylaluminiumhydrid
Formel: C₈H₁₉Al
MW: 142.22 g/mol
Melting Pt: –70 °C
Dichte: 0.701 g/cm³
Flash-Pt: –23 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008928
CAS-Nummer: 1191-15-7
EINECS: 214-729-9
UN: 3394
ADR: 4.2 I

Order Now

Spezifikation Testergebnisse

Appearance Clear solution
Strength 19.5 to 20.5 % (DIBAL-H, on Al basis)
Melting point 118 to 123 °C
Loss on drying ≤0.5 %
Titration with HClO4 98 to 102 %

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR