Air admittance valve
Lieferant: VACUUBRAND
20666800
183-1201EA 175 CHF
183-1201
Air admittance valve
Ventile Luft-Regelventile
These valves are compact air admittance valves with small flange. The VB valve is manually operated. On rotating the knurled screw cap the valve disc is lifted off the seating and air enters through the holes. The valve disc is rotatable. Therefore the valve seat seal is prevented against wear during opening and closing.
Leakage rate: 1×10–6 mbar l/s.