Chemie-Membranpumpe und Chemie-Vakuumsysteme mit ATEX-Zulassung
Lieferant: VACUUBRAND
20696930 20696920 20696921 20696931 20696945 22614250
181-1151EA 11920 CHF
181-1151 181-1149 181-1150 181-1152 181-1153 VACU22614250
Chemie-Membranpumpe und Chemie-Vakuumsysteme mit ATEX-Zulassung
Pumpen Vakuum-Membranpumpen
Chemistry diaphragm pumps: Chemistry diaphragm pumps in equipment category 2 (e.g. for Zone 1) with ATEX compliance. These pumps are exceptionally well designed for the avoidance of ignition sources. They have good chemical resistance, are oil-free, have no friction surfaces and the suction chamber is hermetically sealed off from the drive unit compartment. Motor with flameproof enclosure and integrated, self contained overcurrent and overheating protection for direct 230 V/60 Hz single-phase connection. Diaphragm pump heads made of anti-static materials. Chemically resistant:More Product Information