Natriumdihydrogenphosphat ≥99,0% für die HPLC, Fluka™

Lieferant: Honeywell Chemicals

Synonyms: Sodium dihydrogenphosphate (granular), Sodium dihydrogen orthophosphate, Monobasic Sodium Phosphate, Sodium phosphate monobasic

17844-250G 17844-50G
17844-250GEA 523.2 CHF
17844-250G 17844-50G
Natriumdihydrogenphosphat ≥99,0% für die HPLC, Fluka™
Natriumdihydrogenphosphat
Formel: NaH₂PO₄
Melting Pt: 650 °C
Dichte: 2.36 g/cm³
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003527
CAS-Nummer: 7558-80-7
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806

Order Now

Spezifikation Testergebnisse

Assay 99.0 - 101.0 %
Appearance of the solution (c=10%) complying
pH (10 %, 20°C) 4.0 - 4.5
Absorbance at 210 nm Max. 0.1
Absorbance at 260 nm Max. 0.06
Absorbance 280 nm Max. 0.04
Absorbance at 320 nm Max. 0.02
Absorbance 500 nm Max. 0.02


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR