Thymolblau ACS

Lieferant: Thermo Fisher Scientific

Synonyms: Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

16272.04 16272.09 16272.18
16272.04EA 51.3 CHF
16272.04 16272.09 16272.18
Thymolblau ACS
Thymolblau

CAS No.: 76-61-9 Molecular Formula: C27H30O5S Formula Weight: 466.60 Physical Form: Crystalline Melting Point: 223° dec. Solubility: Insoluble in water. Soluble in alcohol, dilute alkali solutions MDL No.: MFCD00005869


pH indicator: 1.2 (red) to 2.8 (yellow) to 9.6 (blue)

Formel: C₂₇H₃₀O₅S
Melting Pt: 223 °C (dec.)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00005869
CAS-Nummer: 76-61-9
EINECS: 200-973-3
Merck Index: 14,9400

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR