Draining racks, Poxygrid®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
H188050000
149-0197EA 462 CHF
149-0197
Draining racks, Poxygrid®, SP Bel-Art
Gestelle Abtropfgestelle
Racks for drying test tubes, beakers, flasks, and other labware. The stand has rubber tipped feet for non-skid use on a tabletop, or wire loops on the back for wall mounting.
  • Three rows of epoxy-coated hairpin-loop supports (14 in all) hold labware with 18 mm int.Ø necks or larger
  • Top row has 12 straight pins 102 mm long to accommodate smaller items
  • Polystyrene tray catches water as the labware drains