Dysprosium(III)acetat Tetrahydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®

Lieferant: Thermo Fisher Scientific

14595.14 14595.22
14595.14EA 145 CHF
14595.14 14595.22
Dysprosium(III)acetat Tetrahydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Dysprosium(III)acetat Tetrahydrat

Purity: 99.9% CAS No.: 15280-55-4 Molecular Formula: Dy(OOCCH)3.4H2O Formula Weight: 411.68 (339.62anhy) Physical Form: Crystalline Aggregates Odour: Odourless MDL No.: MFCD00013033

Formel: C₆H₉DyO₆·4H₂O
MW: 411.68 g/mol
Melting Pt: 120 °C (Zers.)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00013033
CAS-Nummer: 15280-55-4
EINECS: 242-565-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR