1-Methylpiperazin, Supelco®

Lieferant: Merck


M-Clarity™ quality level = MQ100

14549-5ML-F
14549-5ML-FEA 116 CHF
14549-5ML-F
1-Methylpiperazin, Supelco®
1-Methylpiperazin
Formel: C₅H₁₂N₂
Siedepunkt: 138 °C (1013 hPa)
Melting Pt: –6 °C
Dichte: 0.9085 g/cm³ (20 °C)
Flash-Pt: 34 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00005966
CAS-Nummer: 109-01-3
EINECS: 203-639-5
UN: 2734
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6cf6c0bcba7ca449da26b097658d4ad8c345bfd2&vid=13a0431a4432591788fcfcf1f1e0983a196956fe&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR