Ultrasonic baths, SONOREX TECHNIK
Lieferant: BANDELIN-ELECTRONIC
8200 8210 8220 8230 8250 8270 8240
142-1405EA 3180 CHF
142-1405 142-1410 142-1415 142-1420 142-1425 142-1430 BAND8240
Ultrasonic baths, SONOREX TECHNIK
Ultraschallbäder
Welded cleaning tank made of stainless steel. Floating contamination like particles, oil and grease can be removed from the bath surface using an oil separator.
  • Filling level mark for minimum filling level of the cleaning fluid
  • Additional outlet for connection of an oil separator or for emptying the fluid
  • On/off heat switch heats from 30 to 80°C
  • Integrated ultrasonic generator with 40 kHz frequency
It features 3-way ball valve drain for emptying or refilling the tank or connecting to a filtration.
Additionally, the units from RM 110 UH to RM 210 UHMore Product Information