Titan ≥99,99% (Metall-Basis), Folie, Dicke 0.127 mm (0.005 in)

Lieferant: Thermo Fisher Scientific
Achtung

13976.FF 13976.FI 13976.GH
13976.FFEA 126 CHF
13976.FF 13976.FI 13976.GH
Titan ≥99,99% (Metall-Basis), Folie, Dicke 0.127 mm (0.005 in)
Titanium

Purity: 99.99+% CAS No.: 7440-32-6 Physical Form: ≈0.36g/25x25mm Odour: Odourless MDL No.: MFCD00011264

Formel: Ti
Siedepunkt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Dichte: 4.51 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011264
CAS-Nummer: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR