Lithiumchlorid, ultra trocken ≥99,995% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung


Unter Argon in Ampullen verpackt

13684.06 13684.14 13684.22
13684.06EA 162 CHF
13684.06 13684.14 13684.22
Lithiumchlorid, ultra trocken ≥99,995% (Metall-Basis)
Lithium chloride

Purity: 99.995% CAS No.: 7447-41-8 Molecular Formula: LiCl Formula Weight: 42.39 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 2.068 Melting Point: 605° Boiling Point: 1325-1360° Merck Reference: 14,5528 MDL No.: MFCD00011078

Formel: LiCl
MW: 42.39 g/mol
Siedepunkt: 1325…1360 °C
Melting Pt: 605 °C
Dichte: 2.068
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011078
CAS-Nummer: 7447-41-8
EINECS: 231-212-3
Merck Index: 14,5528

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR