Lithiummolybdat(VI) 99+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Molybdic acid lithium salt, Lithium molybdenum oxide, Lithium molybdate, di-Lithium molybdate

13427.22
13427.22EA 137 CHF
13427.22
Lithiummolybdat(VI) 99+%
Lithiummolybdat(VI)

Purity: 99+% CAS No.: 13568-40-6 Molecular Formula: Li2MoO4 Formula Weight: 173.83 Physical Form: Powder Density: 2.66 Appearance: White Odour: Odourless Melting Point: 705° MDL No.: MFCD00016178

Formel: Li₂MoO₄
MW: 173.82 g/mol
Dichte: 2.66
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00016178
CAS-Nummer: 13568-40-6
EINECS: 236-977-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR