Proflavin Hydrochlorid (3,6-Diaminoacridin Hydrochlorid) 95% (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyms: Acridine-3,6-diamine hydrochloride

131105-25G
131105-25GEA 479 CHF
131105-25G
Proflavin Hydrochlorid (3,6-Diaminoacridin Hydrochlorid) 95% (Farbstoffgehalt), Sigma-Aldrich®
Proflavin Hydrochlorid (3,6-Diaminoacridin Hydrochlorid)

3, 6-Diaminoacridine hydrochloride has been used for studying cation transfer at the liquid–liquid interface. An intercalating dye.

Formel: C₁₃H₁₁N₃·HCl
Melting Pt: 270 °C (dec., lit.)
Lager Temperatur: Room Temperature
MDL Number: MFCD00066867
CAS-Nummer: 952-23-8
EINECS: 213-459-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b449a3ae30fa6ffdc4039c138e1d792a588c5cbf&vid=b449a3ae30fa6ffdc4039c138e1d792a588c5cbf&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance Dark orange to orange-brown powder
Infrared spectrum Conforms to structure.
Uv-visible spectrum In acidified methanol
Carbon 59.7%-67.3%
Nitrogen 15.9%-18.1%
Titration Report % Cl (with silver nitrate)
High pressure liquid 94.0% (minimum)
Solubility 1 mg/ml, H2O; clear, orange to brown


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR