Praseodymtrinitrat Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Praseodymium nitrate hydrate, Nitricacid, praseodymium(3+) salt (3:1), Praseodymium trinitrate hydrate

12909.09 12909.18 12909.30 12909.A1
12909.09EA 69.6 CHF
12909.09 12909.18 12909.30 12909.A1
Praseodymtrinitrat Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Praseodymtrinitrat Hydrat

Praseodymium(III) nitrate hydrate is used in fluorescent display tubes and phosphors. It is also used in the ultrasonic synthesis of phase-pure praseodymium molybdate. It plays a major role in the solvothermal process for formation of doped lanthanide oxysulfide


Purity: 99.9% CAS No.: 15878-77-0 Molecular Formula: Pr(NO3)3.xH2O Formula Weight: 326.92(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Pale green Odour: Odourless UN No.: 1477 MDL No.: MFCD00011175

Formel: N₃O₉Pr
MW: 326.92 g/mol (anhy)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011175
CAS-Nummer: 15878-77-0
EINECS: 233-796-5
UN: 1477
ADR: 5.1 III

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR