Ytterbium(III) nitrate hydrate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Ytterbium trinitrate hydrate

12901.09 12901.18
12901.09EA 157 CHF
12901.09 12901.18
Ytterbium(III) nitrate hydrate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Ytterbium(III) nitrate hydrate

Purity: 99.9% Molecular Formula: Yb(NO3)3.xH2O Formula Weight: 359.05(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White UN No.: UN1477 MDL No.: MFCD00149883

Formel: Yb(NO₃)₃·xH₂O
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149883
EINECS: 237-384-6
UN: 1477
ADR: 5.1,III
Merck Index: 14,10105

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR