TruCLEAN Pro® cleaning system and mops, Perfex
Lieferant: PERFEX
22-38 22-57 8612 22-2W 22-3B 22-33 22-2B 22-2R 22-39 22-3R 22-3W 22-57R 22-58 22-34
129-0101EA 457 CHF
129-0101 129-0130 129-0123 129-0116 129-0118 PERF22-33 129-0115 129-0114 129-0125 129-0117 129-0119 129-0126 129-0100 PERF22-34
TruCLEAN Pro® cleaning system and mops, Perfex
Mopps
TruCLEAN Pro® cleaning systems work to capture and isolate contaminants, ensuring the delivery of unadulterated cleaning and sanitizing agents. TruCLEAN components are constructed of high-grade stainless steel, are easy to maintain, and guaranteed to deliver reliable cleaning results time after time.
  • Reduce the risk of cross-contamination
  • 100% compatible with gamma, ETO autoclave sterilisation
  • Avoid run-off contaminants, common with steam-sterilisers
TruCLEAN Pro® mops:
Designed specifically for use in a cleanroom or sterile environment, TruCLEAN Pro®More Product Information