Triethylamin ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: (Diethylamino)ethane, TEA

12391.30 12391.A1 12391.D5
12391.30EA 106 CHF
12391.30 12391.A1 12391.D5
Triethylamin ≥99%
Triethylamin

Purity: 99+% CAS No.: 121-44-8 Molecular Formula: C6H15N Formula Weight: 101.19 Flash Point: -11°(12°F) Physical Form: Liquid Density: 0.726 Refractive Index: 1.4005 Appearance: Colourless Odour: Ammonia-like Melting Point: -115° Boiling Point: 89-90° Beilstein Reference: 1843166 Merck Reference: 14,9666 UN No.: 1296 MDL No.: MFCD00009051

Formel: (C₂H₅)₃N
Siedepunkt: 89…90 °C
Melting Pt: –115 °C
Dichte: 0.726
Flash-Pt: –11 °C (–12 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00009051
CAS-Nummer: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3,II
Merck Index: 14,9666
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=1c68408a0eedf1bfdb02d1b1e2337d50d449cf9d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR