Kaliumtetrachloroaurat Hydrat ≥99,99% (Metall-Basis) Au ≥ 49%, Premion®

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Gold potassium chloride hydrate, Potassium tetrachloroaurate(III) hydrate, Potassium gold(III) chloride hydrate

12150.03 12150.06
12150.03EA 284 CHF
12150.03 12150.06
Kaliumtetrachloroaurat Hydrat ≥99,99% (Metall-Basis) Au ≥ 49%, Premion®
Kaliumtetrachloroaurat Hydrat

Purity: 99.99% CAS No.: 27988-75-6 Molecular Formula: AuCl4K.xH2O Formula Weight: 377.88(anhy) Physical Form: Crystalline Appearance: Yellow to orange Melting Point: 357° dec. Solubility: Soluble in water Merck Reference: 14,7681 UN No.: 3260 MDL No.: MFCD00043088

Formel: AuCl₄K·xH₂O
MW: 377.88 g/mol (anhy)
Melting Pt: 357 °C (Zers.)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00043088
CAS-Nummer: 27988-75-6
EINECS: 237-190-1
UN: 3260
ADR: 8 III
Merck Index: 14,7681

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR