PIG® spill kit, 115 L overpack
Lieferant: NEW PIG
KIT236 KIT336 KIT436 RFL236 RFL336 RFL436
121-0554EA 374 CHF
121-0554 121-0055 121-0058 121-0555 121-0113 121-0115
PIG® spill kit, 115 L overpack
Absorptionsmaterial
Strong enough to earn a UN rating for transporting spill cleanup waste.
  • Comply with spill plan regs, avoid fines and be ready to respond
  • Overpack drum container is UN-rated (UN1H2/X70/S) for transporting waste after spill cleanup
  • Neatly prepacked container speeds access during a spill emergency
  • Overpack is X-rated in Packing Groups I, II and III for transporting spill cleanup waste by land, sea or rail
  • Lightweight polypropylene container resists chemicals and keeps contents clean and dry
  • Two broom holes 180-degrees opposite each other on the lid allow 100%
More Product Information