Nickel ≥99,994% (Metall-Basis), Folie, Puratronic®, Dicke 0.25 mm (0.01 in)

Lieferant: Thermo Fisher Scientific
Gefahr

12045.FY 12045.HL
12045.FYEA 984 CHF
12045.FY 12045.HL
Nickel ≥99,994% (Metall-Basis), Folie, Puratronic®, Dicke 0.25 mm (0.01 in)
Nickel

Purity: 99.994% CAS No.: 7440-02-0 Physical Form: ≈2.8g/25x50mm Odour: Odourless MDL No.: MFCD00011137

Formel: Ni
MW: 58.69 g/mol
Siedepunkt: 3007
Melting Pt: 1453
Dichte: 8.9 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011137
CAS-Nummer: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR