Natriumdihydrogenphosphat Monohydrat 98,0-102,0 ACS

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate

11591.30 11591.A1 11591.A7
11591.30EA 78.8 CHF
11591.30 11591.A1 11591.A7
Natriumdihydrogenphosphat Monohydrat 98,0-102,0 ACS
Natriumdihydrogenphosphat Monohydrat

Purity: 98.0-102.0% CAS No.: 10049-21-5 Molecular Formula: NaH2PO4.H2O Formula Weight: 137.99 (119.98anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Merck Reference: 14,8660 MDL No.: MFCD00149208

Formel: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Melting Pt: 100 °C (-H₂O)
Dichte: 2.04 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149208
CAS-Nummer: 10049-21-5
EINECS: 231-449-2
Merck Index: 14,8660
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR