Rhodium, Folie, Dicke 0.025mm (0.001 in)

Lieferant: Thermo Fisher Scientific

11524.FF 11524.FI
11524.FFEA 2470 CHF
11524.FF 11524.FI
Rhodium, Folie, Dicke 0.025mm (0.001 in)
Rhodium
Formel: Rh
MW: 102.9 g/mol
Siedepunkt: 3400…3700 °C (1013 hPa)
Melting Pt: 1960
Dichte: 12.41 g/cm³ (20 °C)
MDL Number: MFCD00011201
CAS-Nummer: 7440-16-6
EINECS: 231-125-0
Merck Index: 12,08353

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR