Nickel ≥99,995% (Metall-Basis), Stäbe, Puratronic®, Ø 12 mm (0.47 in)

Lieferant: Thermo Fisher Scientific
Gefahr

11499.HF
11499.HFEA 142 CHF
11499.HF
Nickel ≥99,995% (Metall-Basis), Stäbe, Puratronic®, Ø 12 mm (0.47 in)
Nickel

Purity: 99.995% CAS No.: 7440-02-0 Physical Form: ≈10.1g/cm Odour: Odourless MDL No.: MFCD00011137

Formel: Ni
Siedepunkt: 3007 °C (1013 hPa)
Melting Pt: 1453 °C
Dichte: 8.9 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011137
CAS-Nummer: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR